4-(Pyridin-3-ylsulfonyl)morpholine 98%

Supplier: Ambeed
Warning

A300627-1G A300627-250MG A300627-5G
76875-658EA 61.54 USD
76875-658 76875-660 76875-662
4-(Pyridin-3-ylsulfonyl)morpholine 98%
4-(Pyridin-3-ylsulfonyl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃S
MDL Number: MFCD00090297
CAS Number: 26103-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR