2-(4-Chlorophenoxy)isobutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid

A299870-100G A299870-25G A299870-5G A299870-100MG
76875-648EA 55 USD
76875-648 76875-650 76875-652 77883-821
2-(4-Chlorophenoxy)isobutyric acid 98%
2-(4-Chlorophenoxy)isobutyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.65 g/mol
MDL Number: MFCD00004192
CAS Number: 882-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR