Irsogladine maleate 98%

Supplier: Ambeed
Warning

Synonyms: 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine maleate, 2,4-Diamino-6-(2,5-dichlorophenyl)-1,3,5-triazine Maleate

A299639-100MG A299639-1MG A299639-250MG A299639-25MG A299639-1G A299639-50MG
76875-636EA 21 USD
76875-636 76875-638 76875-640 76875-642 77883-815 77883-816
Irsogladine maleate 98%
Irsogladine maleate

Precaución: for Research Use Only

Formula: C₉H₇Cl₂N₅
MW: 372.16 g/mol
MDL Number: MFCD00873566
CAS Number: 84504-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR