2-(3-Bromo-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A297206-100MG A297206-1G A297206-250MG
76875-612EA 36.08 USD
76875-612 76875-614 76875-616
2-(3-Bromo-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-Bromo-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₂BBrO₃
MW: 353.05 g/mol
MDL Number: MFCD12546588
CAS Number: 1218789-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR