7-Fluoro-1,3-dihydroquinazoline-2,4-dione 98%

Supplier: Ambeed
Warning

A296916-1G A296916-250MG
76875-608EA 24 USD
76875-608 76875-610
7-Fluoro-1,3-dihydroquinazoline-2,4-dione 98%
7-Fluoro-1,3-dihydroquinazoline-2,4-dione

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MDL Number: MFCD09954843
CAS Number: 76088-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR