4-(2-Bromophenyl)morpholine 97%

Supplier: Ambeed
Warning

A295935-100MG A295935-1G A295935-250MG
76875-576EA 8 USD
76875-576 76875-578 76875-580
4-(2-Bromophenyl)morpholine 97%
4-(2-Bromophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD03086182
CAS Number: 87698-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR