Ethyl-3-bromo-6-chloropicolinate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromo-6-chloropicolinate

A295047-1G A295047-250MG A295047-25G A295047-5G
76875-568EA 42 USD
76875-568 76875-570 76875-572 76875-574
Ethyl-3-bromo-6-chloropicolinate 96%
Ethyl 3-bromo-6-chloropicolinate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MDL Number: MFCD11053875
CAS Number: 1065074-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR