5-Norbornene-2-methylamine (mixture of isomers) 97%

Supplier: Ambeed
Danger

Synonyms: 5-Norbornene-2-methylamine, 5-(Aminomethyl)bicyclo[2.2.1]hept-2-ene, 5-(Aminomethyl)norbornene, 5-Norbornene-2-methanamine

A294964-1G A294964-25G A294964-5G A294964-10G
76875-562EA 22.28 USD
76875-562 76875-564 76875-566 77550-790
5-Norbornene-2-methylamine (mixture of isomers) 97%
5-Norbornene-2-methylamine (mixture of isomers)

Precaución: for Research Use Only

Formula: C₈H₁₃N
MW: 123.20 g/mol
MDL Number: MFCD07369043
CAS Number: 95-10-3
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR