3-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperidine-3-carboxylate

A294357-100MG A294357-1G A294357-250MG
76875-540EA 37 USD
76875-540 76875-542 76875-544
3-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 95%
3-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₃₀BNO₅
MDL Number: MFCD09266178
CAS Number: 1073355-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR