6-Ethoxy-2-naphthoic acid 95+%

Supplier: Ambeed
Warning

A293989-100MG A293989-1G A293989-250MG
76875-526EA 6 USD
76875-526 76875-528 76875-530
6-Ethoxy-2-naphthoic acid 95+%
6-Ethoxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
MDL Number: MFCD18815314
CAS Number: 614754-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR