(E)-4-(Dimethylamino)but-3-en-2-one 95%

Supplier: Ambeed
Warning

A293175-100MG A293175-10G A293175-1G A293175-250MG A293175-5G A293175-25G
76875-506EA 5 USD
76875-506 76875-508 76875-510 76875-512 76875-514 77593-792
(E)-4-(Dimethylamino)but-3-en-2-one 95%
(E)-4-(Dimethylamino)but-3-en-2-one

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MDL Number: MFCD00142611
CAS Number: 2802-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR