7-Methyl-1H-indol-4-ol 95%

Supplier: Ambeed
Warning

A292113-100MG A292113-1G A292113-50MG A292113-250MG
76875-492EA 201.58 USD
76875-492 76875-494 76875-496 77612-684
7-Methyl-1H-indol-4-ol 95%
7-Methyl-1H-indol-4-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.17 g/mol
MDL Number: MFCD19218337
CAS Number: 19499-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR