Methyl 4-bromo-5-fluoro-2-hydroxybenzoate 95%
Supplier: Ambeed
|
Warning
|
A291667-100MG
A291667-10G
A291667-1G
A291667-250MG
A291667-5G
76875-474EA
33.95
USD
76875-474
76875-476
76875-478
76875-480
76875-482
Methyl 4-bromo-5-fluoro-2-hydroxybenzoate 95%
Methyl 4-bromo-5-fluoro-2-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₃ MW: 249.034 g/mol |
MDL Number:
MFCD23701539 CAS Number: 1193162-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...