Ethyl-8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

A290890-1G A290890-250MG
76875-456EA 114.58 USD
76875-456 76875-458
Ethyl-8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₂
MW: 283.1212 g/mol
MDL Number: MFCD09475853
CAS Number: 135995-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR