(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Boc-2,2-dimethyloxazolidine-4-carboxylic Acid

A290801-1G A290801-250MG A290801-5G
76875-444EA 26 USD
76875-444 76875-446 76875-448
(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 98%
(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD02682656
CAS Number: 139009-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR