1-(3-Bromo-5-nitrophenyl)ethanone 98%

Supplier: Ambeed
Warning

A290336-100MG A290336-250MG
76875-440EA 37.14 USD
76875-440 76875-442
1-(3-Bromo-5-nitrophenyl)ethanone 98%
1-(3-Bromo-5-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
MDL Number: MFCD20923634
CAS Number: 127413-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR