4-Chloro-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 95%

Supplier: Ambeed
Warning

A290053-100MG A290053-250MG A290053-5G
76875-434EA 100.79 USD
76875-434 76875-436 76875-438
4-Chloro-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 95%
4-Chloro-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD11847536
CAS Number: 582300-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR