3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-2-((2-chlorobenzyl)oxy)-5-methylphenyl)boronic acid

A289689-100MG A289689-1G A289689-250MG
76875-422EA 28.65 USD
76875-422 76875-424 76875-426
3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid 95%
3-Bromo-2-(2-chlorobenzyloxy)-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BBrClO₃
MW: 355.42 g/mol
MDL Number: MFCD06411364
CAS Number: 849052-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR