(3-(((Benzyloxy)carbonyl)amino)-4-fluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A289504-100MG A289504-1G A289504-250MG
76875-416EA 39 USD
76875-416 76875-418 76875-420
(3-(((Benzyloxy)carbonyl)amino)-4-fluorophenyl)boronic acid 98%
(3-(((Benzyloxy)carbonyl)amino)-4-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₄
MDL Number: MFCD19237159
CAS Number: 874219-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR