tert-Butyl (4-bromophenyl)(methyl)carbamate 96%

Supplier: Ambeed
Warning

A288662-1G A288662-250MG A288662-5G
76875-410EA 42 USD
76875-410 76875-412 76875-414
tert-Butyl (4-bromophenyl)(methyl)carbamate 96%
tert-Butyl (4-bromophenyl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MW: 286.16 g/mol
MDL Number: MFCD12913712
CAS Number: 639520-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR