2,4,5-Trichloropyridine 95%

Supplier: Ambeed
Warning

A287059-100MG A287059-1G A287059-250MG A287059-25G A287059-5G
76875-392EA 5 USD
76875-392 76875-394 76875-396 76875-398 76875-400
2,4,5-Trichloropyridine 95%
2,4,5-Trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₃N
MDL Number: MFCD09028037
CAS Number: 55934-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR