4-Bromo-3-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A285718-250MG A285718-1G A285718-5G
76875-380EA 18 USD
76875-380 77568-578 77672-474
4-Bromo-3-hydroxybenzaldehyde 95%
4-Bromo-3-hydroxybenzaldehyde

Precaución: For research use only.

Formula: C₇H₅BrO₂
MDL Number: MFCD11100992
CAS Number: 20035-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR