1,3-Benzoxazole-5-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(1,3-Benzoxazol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole

A285632-100MG A285632-1G A285632-250MG A285632-5G
76875-372EA 33.95 USD
76875-372 76875-374 76875-376 76875-378
1,3-Benzoxazole-5-boronic acid, pinacol ester 97%
1,3-Benzoxazole-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₃
MW: 245.082 g/mol
MDL Number: MFCD13181968
CAS Number: 936902-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR