2-Chloro-5-ethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-5-ethoxyphenyl)boronic acid

A284978-1G A284978-5G
76875-368EA 66.84 USD
76875-368 76875-370
2-Chloro-5-ethoxyphenylboronic acid 98%
2-Chloro-5-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₃
MW: 200.427 g/mol
MDL Number: MFCD08235081
CAS Number: 913835-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR