(2-Methoxypyrimidin-4-yl)methanamine 95%

Supplier: Ambeed
Warning

A284294-100MG A284294-1G A284294-250MG
76875-362EA 41 USD
76875-362 76875-364 76875-366
(2-Methoxypyrimidin-4-yl)methanamine 95%
(2-Methoxypyrimidin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD10697040
CAS Number: 944901-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR