(S)-2-((2-Oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

Synonyms: Rivaroxaban Related Compound G

A283241-10G A283241-1G A283241-250MG A283241-25G A283241-5G A283241-100G
76875-342EA 53.05 USD
76875-342 76875-344 76875-346 76875-348 76875-350 77612-630
(S)-2-((2-Oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)isoindoline-1,3-dione 97%
(S)-2-((2-Oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₂₂H₁₉N₃O₆
MW: 421.40 g/mol
MDL Number: MFCD11977665
CAS Number: 446292-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR