L-(+)-Gulonic acid-γ-lactone 98%

Supplier: Ambeed
Warning

Synonyms: L-Gulonic acid-1,4-lactone, (3S,4R,5R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one, L-Gulonolactone

A282692-100G A282692-1G A282692-25G A282692-500G A282692-5G
76875-332EA 151 USD
76875-332 76875-334 76875-336 76875-338 76875-340
L-(+)-Gulonic acid-γ-lactone 98%
L-(+)-Gulonic acid-gamma-lactone

Precaución: for Research Use Only

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
MDL Number: MFCD00064331
CAS Number: 1128-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR