Methyl-3-methylcrotonate 98% (by GC)

Supplier: Ambeed
Danger

Synonyms: Methyl 3,3-dimethylacrylate, Methyl-3,3-dimethylacrylate

A281313-10G A281313-25G A281313-5G
76875-316EA 6.37 USD
76875-316 76875-318 76875-320
Methyl-3-methylcrotonate 98% (by GC)
Methyl-3-methylcrotonate

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00043940
CAS Number: 924-50-5
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR