LY-333,531 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (7S)-7-((Dimethylamino)methyl)-22,25-dihydro-11H,21H,31H-6-oxa-1,3(3,1)-diindola-2(3,4)-pyrrolacyclononaphane-22,25-dione hydrochloride, LY333531 hydrochloride

A280932-10MG A280932-1MG A280932-5MG
76875-310EA 521.97 USD
76875-310 76875-312 76875-314
LY-333,531 hydrochloride 98%
LY-333,531 hydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₂₈N₄O₃·HCl
MW: 505.1 g/mol
MDL Number: MFCD00940226
CAS Number: 169939-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR