5-Chloro-2,3-dihydro-1H-inden-1-ol 95%
Supplier: Ambeed
|
Warning
|
A280702-100MG
A280702-1G
A280702-250MG
76875-304EA
9
USD
76875-304
76875-306
76875-308
5-Chloro-2,3-dihydro-1H-inden-1-ol 95%
5-Chloro-2,3-dihydro-1H-inden-1-ol
Precaución: for Research Use Only
Formula:
C₉H₉ClO |
MDL Number:
MFCD11219500 CAS Number: 33781-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...