4-Chloro-3-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-methyl-2-nitrobenzene

A280215-10G A280215-25G A280215-500G A280215-5G
76875-278EA 6 USD
76875-278 76875-280 76875-282 76875-284
4-Chloro-3-nitrotoluene 98%
4-Chloro-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 262
Melting Pt: 7
MDL Number: MFCD00007085
CAS Number: 89-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR