5,6-Dibromo-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A279047-100MG A279047-1G A279047-250MG A279047-5G
76875-270EA 19.1 USD
76875-270 76875-272 76875-274 76875-276
5,6-Dibromo-1H-benzo[d]imidazole 97%
5,6-Dibromo-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
MW: 275.93 g/mol
MDL Number: MFCD00795592
CAS Number: 74545-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR