Methyl 2,3-diamino-6-chlorobenzoate 95+%

Supplier: Ambeed
Warning

A278957-100MG A278957-250MG A278957-50MG
76875-264EA 101.85 USD
76875-264 76875-266 76875-268
Methyl 2,3-diamino-6-chlorobenzoate 95+%
Methyl 2,3-diamino-6-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.62 g/mol
MDL Number: MFCD22581571
CAS Number: 635317-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR