2-Bromo-1-chloro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A278509-1G A278509-25G A278509-5G
76875-250EA 24.41 USD
76875-250 76875-252 76875-254
2-Bromo-1-chloro-4-iodobenzene 98%
2-Bromo-1-chloro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl
MW: 317.3495 g/mol
MDL Number: MFCD07780667
CAS Number: 31928-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR