2-((1-Phenoxypropan-2-yl)amino)ethanol 95%

Supplier: Ambeed
Warning

A278492-1G A278492-5G
76875-246EA 8.49 USD
76875-246 76875-248
2-((1-Phenoxypropan-2-yl)amino)ethanol 95%
2-((1-Phenoxypropan-2-yl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
MW: 195.25 g/mol
MDL Number: MFCD00713871
CAS Number: 103-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR