2-Fluoro-2-methylpropan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A278157-100MG A278157-1G A278157-250MG
76875-236EA 89.12 USD
76875-236 76875-238 76875-240
2-Fluoro-2-methylpropan-1-amine hydrochloride 95%
2-Fluoro-2-methylpropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClFN
MW: 127.58 g/mol
MDL Number: MFCD19207842
CAS Number: 879001-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR