2-Chloro-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Phe(2-Cl)-OH

A278024-10G A278024-1G A278024-250MG A278024-25G A278024-5G
76875-226EA 147.47 USD
76875-226 76875-228 76875-230 76875-232 76875-234
2-Chloro-D-phenylalanine 98%
2-Chloro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD00077920
CAS Number: 80126-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR