Boc-D-Methioninol 98%

Supplier: Ambeed
Warning

A277797-1G A277797-250MG A277797-5G
76875-218EA 30.01 USD
76875-218 76875-220 76875-222
Boc-D-Methioninol 98%
Boc-D-Methioninol
Formula: C₁₀H₂₁NO₃S
MDL Number: MFCD00235940
CAS Number: 91177-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR