3-(4-Fluorophenyl)pentanedioic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluorophenyl)glutaric acid

A276439-100MG A276439-250MG
76875-194EA 19.1 USD
76875-194 76875-196
3-(4-Fluorophenyl)pentanedioic acid 95%
3-(4-Fluorophenyl)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
MW: 226.201 g/mol
MDL Number: MFCD19352347
CAS Number: 3449-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR