JNJ-38877605 98+%

Supplier: Ambeed
Warning

Synonyms: 6-(Difluoro(6-(1-methyl-1H-pyrazol-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl)quinoline

A275907-10MG A275907-1MG A275907-50MG A275907-5MG
76875-184EA 61 USD
76875-184 76875-186 76875-188 76875-190
JNJ-38877605 98+%
JNJ-38877605

Precaución: for Research Use Only

Formula: C₁₉H₁₃F₂N₇
MDL Number: MFCD11977274
CAS Number: 943540-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR