5-Amino-3-methylisoxazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
A274934-1G
A274934-250MG
A274934-25G
A274934-5G
76875-170EA
6.37
USD
76875-170
76875-172
76875-174
76875-176
5-Amino-3-methylisoxazole 97%
5-Amino-3-methylisoxazole
Precaución: for Research Use Only
Formula:
C₄H₆N₂O MW: 98.1 g/mol |
MDL Number:
MFCD00003151 CAS Number: 14678-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...