Methyl morpholine-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A273902-1G A273902-250MG A273902-5G
76875-148EA 115.64 USD
76875-148 76875-150 76875-152
Methyl morpholine-3-carboxylate hydrochloride 97%
Methyl morpholine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃·ClH
MW: 181.617 g/mol
MDL Number: MFCD09263285
CAS Number: 1214686-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR