Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate 97%

Supplier: Ambeed
Danger

A273497-100G A273497-1G A273497-250MG A273497-5G A273497-25G A273497-10G
76875-140EA 304 USD
76875-140 76875-142 76875-144 76875-146 77593-686 77612-592
Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate 97%
Ethyl 2-chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD02852962
CAS Number: 27143-07-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR