5,6-Diamino-4-hydroxy-2-mercaptopyrimidine 85%

Supplier: Ambeed
Warning

A273422-10G A273422-1G A273422-25G A273422-5G
76875-132EA 62.6 USD
76875-132 76875-134 76875-136 76875-138
5,6-Diamino-4-hydroxy-2-mercaptopyrimidine 85%
5,6-Diamino-4-hydroxy-2-mercaptopyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄OS
MW: 158.18 g/mol
MDL Number: MFCD00040963
CAS Number: 1004-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR