3,3-Dimethylbenzo[c][1,2]oxaborol-1(3H)-ol 95%

Supplier: Ambeed
Warning

A273232-100MG A273232-1G A273232-250MG A273232-25G
76875-124EA 36 USD
76875-124 76875-126 76875-128 77883-756
3,3-Dimethylbenzo[c][1,2]oxaborol-1(3H)-ol 95%
3,3-Dimethylbenzo[c][1,2]oxaborol-1(3H)-ol

Precaución: for Research Use Only

Formula: C₉H₁₁BO₂
MDL Number: MFCD13180474
CAS Number: 221352-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR