(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluoro-1,3-benzodioxole-5-boronic acid, (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

A272980-1G A272980-250MG A272980-5G
76875-114EA 135.8 USD
76875-114 76875-116 76875-118
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
MDL Number: MFCD08706236
CAS Number: 190903-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR