1H-Benzimidazole-2-carboxylic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-benzo[d]imidazole-2-carboxylate

A272959-1G A272959-250MG A272959-25G A272959-5G
76875-106EA 40.32 USD
76875-106 76875-108 76875-110 76875-112
1H-Benzimidazole-2-carboxylic acid methyl ester 98%
1H-Benzimidazole-2-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
MDL Number: MFCD03413579
CAS Number: 5805-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR