5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-(Trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

A272514-250MG A272514-100MG
76875-098EA 87 USD
76875-098 77672-406
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 95%
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₄H₄BF₃N₂O₂
MW: 179.893 g/mol
MDL Number: MFCD07375369
CAS Number: 1202054-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR