3-(3,5-Dichlorophenyl)acrylic acid 95%

Supplier: Ambeed
Warning

A272500-1G A272500-250MG
76875-094EA 86 USD
76875-094 76875-096
3-(3,5-Dichlorophenyl)acrylic acid 95%
3-(3,5-Dichlorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₂
MDL Number: MFCD00179444
CAS Number: 90418-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR