Ramipril 98%

Supplier: Ambeed
Danger

Synonyms: (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-cyclopenta[b]pyrrole-2-carboxylic acid, Altace, HOE498, (2S,3aS,6aS)-1-((S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoyl)octahydrocyclopenta[b]pyrrole-2-carboxylic acid

A271148-100MG A271148-1G A271148-25G A271148-5G
76875-082EA 5.31 USD
76875-082 76875-084 76875-086 76875-088
Ramipril 98%
Ramipril

Precaución: for Research Use Only

Formula: C₂₃H₃₂N2₂O₅
MW: 416.51 g/mol
MDL Number: MFCD00865775
CAS Number: 87333-19-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR